ENA333557367 Enamine

3-bromopyridine-2-thiol

  • MDL number MFCD14651478
  • Cas Number : 65938-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80846
Availability details for
Quantity
  • Availability
  • Add to Cart