ENA333557384 Enamine

4-amino-5-[3-(dimethylamino)phenyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD11186283
  • Cas Number : 1094226-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80880
Availability details for
Quantity
  • Availability
  • Add to Cart