ENA333557387 Enamine

2-(3-chloro-2-methylphenoxy)acetic acid

  • MDL number MFCD19686263
  • Cas Number : 579-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80898

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80898
Availability details for
Quantity
  • Availability
  • Add to Cart