ENA333557393 Enamine

2-(4-hydroxyphenyl)-N-(piperidin-4-yl)acetamide

  • MDL number MFCD11642769
  • Cas Number : 1097827-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80905
Availability details for
Quantity
  • Availability
  • Add to Cart