ENA333557403 Enamine

3-(pyridin-2-yl)-4,5-dihydro-1,2,4-oxadiazole-5-thione

  • MDL number MFCD17259801
  • Cas Number : 1334147-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80936

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80936
Availability details for
Quantity
  • Availability
  • Add to Cart