ENA333557410 Enamine

dimethyl({2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethyl})amine

  • MDL number MFCD09746202
  • Cas Number : 925940-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80943
Availability details for
Quantity
  • Availability
  • Add to Cart