ENA333557411 Enamine

1H,5H-pyrido[2,3-d]pyridazin-5-one

  • MDL number MFCD19159813
  • Cas Number : 1314978-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80961
Availability details for
Quantity
  • Availability
  • Add to Cart