ENA333557417 Enamine

6-bromoimidazo[1,2-a]pyridine-3-carbaldehyde

  • MDL number MFCD09864909
  • Cas Number : 30384-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80970
Availability details for
Quantity
  • Availability
  • Add to Cart