ENA333557419 Enamine

2-(5-methyl-1H-1,2,4-triazol-3-yl)propan-2-ol

  • MDL number MFCD19686271
  • Cas Number : 101822-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80973
Availability details for
Quantity
  • Availability
  • Add to Cart