ENA333557426 Enamine

2-phenylazepane

  • MDL number MFCD02663698
  • Cas Number : 3466-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58298
Availability details for
Quantity
  • Availability
  • Add to Cart