ENA333557428 Enamine

2-(3-fluorophenyl)-2-methylpropanoic acid

  • MDL number MFCD11036911
  • Cas Number : 93748-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80987

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80987
Availability details for
Quantity
  • Availability
  • Add to Cart