ENA333557430 Enamine

ethyl 1-benzyl-3-hydroxypyrrolidine-3-carboxylate

  • MDL number MFCD17317132
  • Cas Number : 1274104-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80989
Availability details for
Quantity
  • Availability
  • Add to Cart