ENA333557433 Enamine

4-(1H-pyrazol-1-yl)pyridine-2-carboxylic acid

  • MDL number MFCD11181367
  • Cas Number : 1152523-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80995

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80995
Availability details for
Quantity
  • Availability
  • Add to Cart