ENA333557436 Enamine

1-(3-methylphenyl)ethan-1-ol

  • MDL number MFCD00046635
  • Cas Number : 7287-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80998

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-80998
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart