ENA333557441 Enamine

5,7-dimethyl-1,8-naphthyridin-2-amine

  • MDL number MFCD00006732
  • Cas Number : 39565-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81013
Availability details for
Quantity
  • Availability
  • Add to Cart