ENA333557445 Enamine

4-(2-oxo-1,2-dihydropyridin-1-yl)butanenitrile

  • MDL number MFCD09947046
  • Cas Number : 218921-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59021
Availability details for
Quantity
  • Availability
  • Add to Cart