ENA333557448 Enamine

2-methyl-1-(3-nitro-1H-pyrazol-1-yl)propan-2-ol

  • MDL number MFCD14594920
  • Cas Number : 1003013-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81025
Availability details for
Quantity
  • Availability
  • Add to Cart