ENA333557457 Enamine

3-fluoro-4-(4-methyl-1H-pyrazol-1-yl)aniline

  • MDL number MFCD08699596
  • Cas Number : 1006960-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81035
Availability details for
Quantity
  • Availability
  • Add to Cart