ENA333557461 Enamine

1-(2,2-dimethylpropanoyl)-2,3-dihydro-1H-indole-5-carboxylic acid

  • MDL number MFCD11539106
  • Cas Number : 1097098-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81042
Availability details for
Quantity
  • Availability
  • Add to Cart