ENA333557464 Enamine

(2-isocyanatocyclopropyl)benzene

  • MDL number MFCD16300991
  • Cas Number : 63006-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81047
Availability details for
Quantity
  • Availability
  • Add to Cart