ENA333557470 Enamine

3-methyl-4-[2-(pyrrolidin-1-yl)ethoxy]aniline

  • MDL number MFCD12786493
  • Cas Number : 862874-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81284

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81284
Availability details for
Quantity
  • Availability
  • Add to Cart