ENA333557471 Enamine

1-benzyl-5-methoxy-2-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD00409375
  • Cas Number : 132105-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81285

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81285
Availability details for
Quantity
  • Availability
  • Add to Cart