ENA333557483 Enamine

1,4-dioxa-8-azaspiro[4.6]undecan-9-one

  • MDL number MFCD16657119
  • Cas Number : 172090-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81419

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81419
Availability details for
Quantity
  • Availability
  • Add to Cart