ENA333557493 Enamine

propan-2-yl (2S)-pyrrolidine-2-carboxylate

  • MDL number MFCD00191005
  • Cas Number : 61114-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81491
Availability details for
Quantity
  • Availability
  • Add to Cart