ENA333557497 Enamine

methyl 2-{[2-(2,4-dichlorophenyl)ethyl]amino}acetate

  • MDL number MFCD11152574
  • Cas Number : 1021120-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81495
Availability details for
Quantity
  • Availability
  • Add to Cart