ENA333557506 Enamine

2-chloro-4,6-dimethylpyridin-3-amine

  • MDL number MFCD08273051
  • Cas Number : 140413-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81518
Availability details for
Quantity
  • Availability
  • Add to Cart