ENA333557512 Enamine

2-(methylamino)-5-nitropyridine-3-carbonitrile

  • MDL number MFCD16661607
  • Cas Number : 31309-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81527
Availability details for
Quantity
  • Availability
  • Add to Cart