ENA333557514 Enamine

2-[(4-fluorophenyl)methoxy]phenol

  • MDL number MFCD11181832
  • Cas Number : 1094248-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81531
Availability details for
Quantity
  • Availability
  • Add to Cart