ENA333557519 Enamine

4,4-dimethylpent-1-en-3-one

  • MDL number MFCD18072789
  • Cas Number : 2177-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60338
Availability details for
Quantity
  • Availability
  • Add to Cart