ENA333557525 Enamine

(2S,4S)-1-[(tert-butoxy)carbonyl]-4-(trifluoromethyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08460520
  • Cas Number : 470482-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-127
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60590
Availability details for
Quantity
  • Availability
  • Add to Cart