ENA333557540 Enamine

5-fluoro-N1-(propan-2-yl)benzene-1,2-diamine

  • MDL number MFCD16744198
  • Cas Number : 1026934-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62567
Availability details for
Quantity
  • Availability
  • Add to Cart