ENA333557542 Enamine

2-amino-5-(tert-butylamino)-4-fluorobenzoic acid

  • MDL number MFCD16159096
  • Cas Number : 1248655-94-8
  • Molecular Weight : 226.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-81584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81584
Availability details for
Quantity
  • Availability
  • Add to Cart