ENA333557543 Enamine

5-amino-2-methanesulfonyl-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid

  • MDL number MFCD18010368
  • Cas Number : 1281581-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81585
Availability details for
Quantity
  • Availability
  • Add to Cart