ENA333557546 Enamine

1-(5-chloro-2-methoxyphenyl)propan-1-one

  • MDL number MFCD11617658
  • Cas Number : 68597-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81588

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81588
Availability details for
Quantity
  • Availability
  • Add to Cart