ENA333557549 Enamine

[1-(4-chlorophenyl)propan-2-yl](methyl)amine

  • MDL number MFCD01708360
  • Cas Number : 1199-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81591

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81591
Availability details for
Quantity
  • Availability
  • Add to Cart