ENA333557550 Enamine

2-(2-ethyl-1H-1,3-benzodiazol-1-yl)ethan-1-amine

  • MDL number MFCD10024204
  • Cas Number : 61981-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63467
Availability details for
Quantity
  • Availability
  • Add to Cart