ENA333557553 Enamine

2-[2-(3-hydroxybenzamido)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD09049311
  • Cas Number : 926207-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64039
Availability details for
Quantity
  • Availability
  • Add to Cart