ENA333557555 Enamine

tert-butyl 4-(2-chloropropanoyl)piperazine-1-carboxylate

  • MDL number MFCD11931565
  • Cas Number : 1181803-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64286
Availability details for
Quantity
  • Availability
  • Add to Cart