ENA333557556 Enamine

3-amino-N-(4-methoxybenzyl)-1H-pyrazole-4-sulfonamide

  • MDL number MFCD12522342
  • Cas Number : 1184583-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81601
Availability details for
Quantity
  • Availability
  • Add to Cart