ENA333557557 Enamine

3-(1H-1,3-benzodiazol-1-yl)-2-(propylamino)propanoic acid dihydrochloride

  • MDL number MFCD19686305
  • Cas Number : 1333875-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81602
Availability details for
Quantity
  • Availability
  • Add to Cart