ENA333557572 Enamine

4-(chloromethyl)-2-(1-ethoxyethyl)-1,3-thiazole

  • MDL number MFCD16079180
  • Cas Number : 1247625-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81622
Availability details for
Quantity
  • Availability
  • Add to Cart