ENA333557575 Enamine

1-(2,4-difluorophenyl)-3-(furan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD08442972
  • Cas Number : 926188-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81625
Availability details for
Quantity
  • Availability
  • Add to Cart