ENA333557582 Enamine

3-(2-amino-1,3-thiazol-4-yl)phenol

  • MDL number MFCD01627944
  • Cas Number : 155983-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81633
Availability details for
Quantity
  • Availability
  • Add to Cart