ENA333557589 Enamine

1-methyl-5-oxo-2-(thiophen-2-yl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD07380190
  • Cas Number : 1248551-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81640
Availability details for
Quantity
  • Availability
  • Add to Cart