ENA333557590 Enamine

methyl 2-(methylamino)-2-phenylacetate

  • MDL number MFCD12148297
  • Cas Number : 107635-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81641
Availability details for
Quantity
  • Availability
  • Add to Cart