ENA333557597 Enamine

3-chloro-2-[(2,2-difluoroethyl)sulfanyl]aniline

  • MDL number MFCD12768620
  • Cas Number : 1179075-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81659
Availability details for
Quantity
  • Availability
  • Add to Cart