ENA333557611 Enamine

N-({2-chloroimidazo[1,2-a]pyridin-3-yl}methylidene)hydroxylamine

  • Cas Number : 1251406-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81679
Availability details for
Quantity
  • Availability
  • Add to Cart