ENA333557613 Enamine

3-chloro-N-[1-(N'-hydroxycarbamimidoyl)-1-methylethyl]benzamide

  • Cas Number : 1286215-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81681
Availability details for
Quantity
  • Availability
  • Add to Cart