ENA333557616 Enamine

ethyl 3-hydroxy-2-nitrobenzoate

  • MDL number MFCD02069076
  • Cas Number : 81914-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81685
Availability details for
Quantity
  • Availability
  • Add to Cart