ENA333557618 Enamine

2-methyl-5-[(morpholin-4-yl)carbonyl]aniline

  • MDL number MFCD09043756
  • Cas Number : 21447-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81687

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81687
Availability details for
Quantity
  • Availability
  • Add to Cart